EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Molecular dynamics of glycine ions in alanine doped TGS single crystal as probed by polarized laser Raman spectroscopy.
PMID 22947647
Abstract
Polarized Raman spectra of pure and alanine doped tri-glycine sulfate (TGS) single crystals at 12 K in different scattering geometries are analyzed. Sub species modes due to three crystallographically distinguishable glycine ions G (I), G (II) and G (III) are assigned. It is observed that alanine doping does not change the crystalline field and acts as local perturbation only. The major changes due to doping are observed in the relative intensities of different modes; most of the modes associated with G (I) and SO(4)(2-) ions show reversal behavior in relative intensity at high doping concentration. The observed spectral changes are analyzed in terms of reorientation of G (I) ions with sub species modes of G (II)/ G (III) following the reorientation due to complex hydrogen bonding network.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G1377
Gly-Gly-Gly
C6H11N3O4
50239
Gly-Gly-Gly, BioUltra, ≥99.0% (NT)
C6H11N3O4