EMAIL THIS PAGE TO A FRIEND
Nanotechnology
Interplay between crystal phase purity and radial growth in InP nanowires.
PMID 22948129
Abstract
The interplay between crystal phase purity and radial growth in InP nanowires is investigated. By modifying the growth rate and V/III ratio, regions of high or low stacking fault density can be controllably introduced into wurtzite nanowires. It is found that regions with high stacking fault density encourage radial growth. Through careful choice of growth conditions pure wurtzite InP nanowires are then grown which exhibit narrow 4.2 K photoluminescence linewidths of 3.7 meV at 1.490 meV, and no evidence of emission related to stacking faults or zincblende insertions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
366870
Indium(III) phosphide, pieces, 3-20 mesh, 99.998% trace metals basis
InP