EMAIL THIS PAGE TO A FRIEND
European neurology
The impact of rotigotine on cardiovascular autonomic function in early Parkinson's disease.
PMID 22948481
Abstract
Dysautonomia can occur in early stages of Parkinson's disease (PD) influencing tolerance to dopaminergic therapies. Rotigotine, a non-ergot dopamine agonist, has recently been developed as an effective alternative antiparkinsonian drug, but its influence on the autonomic nervous system was not investigated. Twenty subjects out of 34 consecutive de novo PD patients were submitted to full assessment of cardiovascular autonomic function before and after reaching a stable rotigotine regimen [6 mg/24 h (n = 3) or 8 mg/24 h (n = 17)]. Patients reached significant clinical improvement (-27% on the Unified Parkinson's Disease Rating Scale part III) and did not show significant differences in cardiovascular tests compared to baseline data. However, an unexpected trend towards increasing systolic blood pressure after head-up tilt test was detected. Our study demonstrates that rotigotine does not influence cardiovascular autonomic responses in early de novo PD patients. Consequently, it may represent a well-tolerated and efficacious therapeutic option in newly diagnosed PD subjects.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R9281
Rotigotine hydrochloride, ≥98% (HPLC)
C19H25NOS·HCl