EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Use of xylitol to enhance the therapeutic efficacy of polymethylmethacrylate-based antibiotic therapy in treatment of chronic osteomyelitis.
PMID 22948866
Abstract
Using a rabbit model of postsurgical osteomyelitis, we demonstrate that incorporation of xylitol into polymethylmethacrylate (PMMA) bone cement enhances the elution of daptomycin under in vivo conditions. We also demonstrate that this can be correlated with an improved therapeutic outcome in the treatment of a chronic bone infection following surgical debridement.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
PHR1280
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material
C5H12O5