EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Silver triflate-catalyzed tandem reaction of N'-(2-alkynylbenzylidene)hydrazide with pyridyne.
PMID 22948904
Abstract
A silver triflate-catalyzed tandem reaction of N'-(2-alkynylbenzylidene)hydrazide with pyridyne is presented. Different outcomes are obtained, depending on the pyridynes utilized in the transformation.