EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
Regulation of immunity and disease resistance by commensal microbes and chromatin modifications during zebrafish development.
PMID 22949679
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-942
Anti-acetyl-Histone H3 (Lys9) Antibody, clone, Upstate®, from rabbit