EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Nitroxide radicals as highly reactive redox mediators in dye-sensitized solar cells.
PMID 22951945
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
745618
D205 Dye, 97% (HPLC)
C48H47N3O4S3