EMAIL THIS PAGE TO A FRIEND
ChemMedChem
Design and synthesis of 3-carbamoylbenzoic acid derivatives as inhibitors of human apurinic/apyrimidinic endonuclease 1 (APE1).
PMID 22952012
Abstract
Apurinic/apyrimidinic (AP) endonuclease 1 (APE1) is a multifaceted protein with an essential role in the base excision repair (BER) pathway. Its implication in tumor development, progression, and resistance has been confirmed in multiple cancers, making it a viable target for intensive investigation. In this work, we designed and synthesized different classes of small-molecule inhibitors of the catalytic endonuclease function of APE1 that contain a 3-carbamoylbenzoic acid scaffold. Further structural modifications were made with the aim of increasing the activity and cytotoxicity of these inhibitors. Several of our compounds were shown to inhibit the catalytic endonuclease function of APE1 with potencies in the low-micromolar range in vitro, and therefore represent novel classes of APE1 inhibitors worthy of further development.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D5412
2′-Deoxyuridine, ≥98.5%
C9H12N2O5
D4879
N,N′-Bis(hydroxymethyl)urea, ≥95%
C3H8N2O3