PloS one
Biosignatures for Parkinson's disease and atypical parkinsonian disorders patients.
PMID 22952715
Abstract
Diagnosis of Parkinson' disease (PD) carries a high misdiagnosis rate due to failure to recognize atypical parkinsonian disorders (APD). Usually by the time of diagnosis greater than 60% of the neurons in the substantia nigra are dead. Therefore, early detection would be beneficial so that therapeutic intervention may be initiated early in the disease process. We used splice variant-specific microarrays to identify mRNAs whose expression is altered in peripheral blood of early-stage PD patients compared to healthy and neurodegenerative disease controls. Quantitative polymerase chain reaction assays were used to validate splice variant transcripts in independent sample sets. Here we report a PD signature used to classify blinded samples with 90% sensitivity and 94% specificity and an APD signature that resulted in a diagnosis with 95% sensitivity and 94% specificity. This study provides the first discriminant functions with coherent diagnostic signatures for PD and APD. Analysis of the PD biomarkers identified a regulatory network with nodes centered on the transcription factors HNF4A and TNF, which have been implicated in insulin regulation.