EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
A new bioactive Schiff base ligands derived from propylazo-N-pyrimidin-2-yl-benzenesulfonamides Mn(II) and Cu(II) complexes: synthesis, thermal and spectroscopic characterization biological studies and 3D modeling structures.
PMID 22954685
Abstract
New series of Schiff base ligand H(2)L and their Cu(II) and Mn(II) complexes derived from azosulfapyrimidine were synthesized and characterized by elemental and thermal studies conductance measurements IR, electronic and EPR spectra. 3D modeling of the ligand indicate that azo group does not participate in complex formation and surface potential on one of the ligand under study indicate that electron density around azomethine groups are much higher than the azo group therefore coordination takes place around azomethine groups. The variety in the geometrical structures depends on the nature of both the metal ions and the Schiff base ligands. The thermo kinetic parameters are calculated and discussed. The biological activities of the ligands and complexes have been screened in vitro against some bacteria and fungi to study their capacity to inhibit their growth and to study the toxicity of the compounds.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
108146
Benzenesulfonamide, ≥98%
C6H7NO2S