PloS one
Immunosuppressive treatment for nephrotic idiopathic membranous nephropathy: a meta-analysis based on Chinese adults.
PMID 22957065
Abstract
Idiopathic membranous nephropathy (IMN) is the most common pathological type for nephrotic syndrome in adults in western countries and China. The benefits and harms of immunosuppressive treatment in IMN remain controversial. To assess the efficacy and safety of different immunosuppressive agents in the treatment of nephrotic syndrome caused by IMN. PubMed, EMBASE, Cochrane Library and wanfang, weipu, qinghuatongfang, were searched for relevant studies published before December 2011. Reference lists of nephrology textbooks, review articles were checked. A meta-analysis of randomized controlled trials (RCTs) meeting the criteria was performed using Review Manager. 17 studies were included, involving 696 patients. Calcineurin inhibitors had a better effect when compared to alkylating agents, on complete remission (RR 1.61, 95% CI 1.13, to 2.30 P = 0.008), partial or complete remission (effective) (CR/PR, RR 1.29, 95% CI 1.09 to 1.52 P = 0.003), and fewer side effects. Among calcineurin inhibitors, tacrolimus (TAC) was shown statistical significance in inducing more remissions. When compared to cyclophosphamide (CTX), leflunomide (LET) showed no beneficial effect, mycophenolate mofetil (MMF) showed significant beneficial on effectiveness (CR/PR, RR: 1.41, 95% CI 1.16 to 1.72 P = 0.0006) but not significant on complete remission (CR, RR: 1.38, 95% CI 0.89 to 2.13 P = 0.15). This analysis based on Chinese adults and short duration RCTs suggested calcineurin inhibitors, especially TAC, were more effective in proteinuria reduction in IMN with acceptable side effects. Long duration RCTs were needed to confirm the long-term effects of those agents in nephrotic IMN.