EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Microfluidic size selective growth of palladium nano-particles on carbon nano-onions.
PMID 22957341
Abstract
Size selective growth of palladium nano-particles 2-7 nm in diameter on the surface of carbon nano-onions (CNOs) (derived from catalytic cracking of methane) in water involves pretreating the CNOs with p-phosphonic acid calix[8]arene then H(2)PdCl(4) followed by dynamic thin film processing under hydrogen in a vortex fluidic device.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
215112
Phosphorous acid, 99%
H3O3P
04115
Phosphorous acid solution, ≥50%
H3O3P