EMAIL THIS PAGE TO A FRIEND
Journal of fish biology
Immersion anaesthesia with tricaine methanesulphonate or propofol on different sizes and strains of silver catfish Rhamdia quelen.
PMID 22957883
Abstract
The efficacy of immersion anaesthesia with tricaine methanesulphonate (MS222) or propofol on albino and grey silver catfish Rhamdia quelen was assessed through induction and recovery times and observation of mortality. Besides reporting a novel, efficient and practical use of propofol as an immersion anaesthetic, the study shows that it is essential to consider size and strain when anaesthetizing R. quelen with MS222 or propofol bath solution in order to minimize physiological impact.