EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Sugar-metal ion interactions: the coordination behaviors of lanthanum with erythritol.
PMID 22960209
Abstract
Three novel lanthanum chloride-erythritol complexes (LaCl(3)·C(4)H(10)O(4)·5H(2)O (LaE(I)), LaCl(3)·C(4)H(10)O(4)·3H(2)O (LaE(II)), and LaCl(3)·1.5C(4)H(10)O(4) (LaE(III)) were synthesized and characterized by single crystal X-ray diffraction, FTIR, far-IR, THz, and Raman spectroscopy. The coordination number of La(3+) is nine. LaE(I) and LaE(II) have similar coordination spheres, but their hydrogen bond networks are different. Erythritol exhibits two coordination modes: two bidentate ligands and tridentate ligands in LaE(III). Chloride ions and water coordinate with La(3+) or participate in the hydrogen-bond networks in the three complexes. Crystal structures, FTIR, FIR, THz, and Raman spectra provide detailed information on the structures and coordination of hydroxyl groups to metal ions in the metal-carbohydrate complexes.