EMAIL THIS PAGE TO A FRIEND
Cold Spring Harbor perspectives in biology
Imaging translation in single cells using fluorescent microscopy.
PMID 22960595
Abstract
The regulation of translation provides a mechanism to control not only the abundance of proteins, but also the precise time and subcellular location that they are synthesized. Much of what is known concerning the molecular basis for translational control has been gleaned from experiments (e.g., luciferase assays and polysome analysis) that measure average changes in the protein synthesis of a population of cells, however, mechanistic insights can be obscured in ensemble measurements. The development of fluorescent microscopy techniques and reagents has allowed translation to be studied within its cellular context. Here we highlight recent methodologies that can be used to study global changes in protein synthesis or regulation of specific mRNAs in single cells. Imaging of translation has provided direct evidence for local translation of mRNAs at synapses in neurons and will become an important tool for studying translational control.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P7255
Puromycin dihydrochloride from Streptomyces alboniger, ≥98% (HPLC), powder
C22H29N7O5 · 2HCl