EMAIL THIS PAGE TO A FRIEND
Chembiochem : a European journal of chemical biology
Mapping the mechanism of the resorcinol ring formation catalyzed by ArsB, a type III polyketide synthase from Azotobacter vinelandii.
PMID 22961888
Abstract
Who's first? Aldol cyclization occurs before hydrolysis in the resorcinol ring formation catalyzed by the type III polyketide synthase, ArsB. Synthetic C(20)-TKA was not converted to alkylresorcinol by ArsB, but rather inhibited the enzyme activity, thus indicating that C(20)-TKA is not an intermediate in ArsB-catalyzed alkylresorcinol formation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
53363
Resorcinol, certified reference material, TraceCERT®
C6H6O2