EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
On the aromatic stabilization of benzenoid hydrocarbons.
PMID 22962659
Abstract
A general scheme for estimation of aromatic stabilization energies of benzenoid hydrocarbons based on selected topological features has been presented. The reactions have been applied to benzene, naphthalene, anthracene, phenanthrene, pyrene, tetracene, benz[a]anthracene, chrysene, [4]-helicene, anthanthrene and coronene.