EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Enantioselectivity in visible light-induced, singlet oxygen [2+4] cycloaddition reactions (type II photooxygenations) of 2-pyridones.
PMID 22962664
Abstract
3-Substituted 2-pyridones were enantioselectively (68-90% ee) converted into the respective 3-hydroxypyridine-2,6-diones by a sequence consisting of a template-mediated type II photooxygenation and an acid-catalysed rearrangement.
Related Materials