EMAIL THIS PAGE TO A FRIEND
Archives of animal nutrition
The effects of iodine level and source on iodine carry-over in eggs and body tissues of laying hens.
PMID 22962946
Abstract
In the presented study the effect of different iodine (I) levels and sources in hen feed on the iodine concentration of different tissues, blood serum, and eggs of laying hens was studied. For this purpose, two experiments were conducted with 30 laying hens each. In these experiments feed was enriched with KI and Ca(IO(3))(2), respectively, at 0 (Control), 0.25, 0.5, 2.5 and 5 mg I/kg feed, resulting a analysed iodine level from 0.44 to 4.20 mg/kg feed. After four weeks experimental feeding the iodine concentrations of thyroid glands, blood, meat, liver, abdominal fat and eggs were measured with inductively coupled plasma mass spectrometry. The experimental treatment did not affect hen performance. The iodine supplementation significantly increased the iodine concentration of eggs (144-1304 μg/kg), thyroid glands (3367-5975 μg/g), blood serum (16-67 μg/kg) and liver (13-43 μg/kg). Meat (about 14 μg I/kg) and abdominal fat (about 12 μg I/kg) were not significantly affected by iodine treatment. Comparative regression analyses showed that at a similar iodine intake, the supply via KI resulted in significantly higher iodine deposition into eggs than Ca(IO(3))(2). Due to the high carry-over of iodine into eggs, eggs may considerably contribute to the iodine supply of the consumers.