EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Enantioselective behavior of malathion enantiomers in toxicity to beneficial organisms and their dissipation in vegetables and crops.
PMID 22964386
Abstract
The dissipation behavior of the two enantiomers of malathion was elucidated in five plant species using enantioselective high performance liquid chromatography (HPLC), and the acute toxicity of the individual enantiomers toward earthworms and honeybees was studied. The calculated LC(50) values of the R-, S- and rac-malathion to earthworms were 0.3869, 25.17, and 19.19 μg/cm(2), respectively, while the calculated LC(50) values of R-, S- and rac-malathion to bees were 2.15, 36.67, and 7.11 μg/mL, respectively. This indicated that the R-enantiomer was more toxic than S-enantiomer. The results of the degradation of racemate in Chinese cabbage and rape showed that the inactive S-(-)-enantiomer degraded faster than the active R-(+)-enantiomer. Inversely, we found a preferential degradation of the R-(+)-enantiomer in sugar beet. However, the degradation of malathion in paddy rice and wheat were nonenantioselectivity. In all plants, malathion was degraded to levels <10% after 5 days, and the calculated t(½) values of the enantiomers ranged from 0.83 to 1.43 days in these five plants. In conclusion, our findings of enantioselectivity in the environmental fate and acute toxicity of the malathion enantiomers may have implications for better environmental and ecological risk assessment for chiral pesticides in general.