EMAIL THIS PAGE TO A FRIEND
JOP : Journal of the pancreas
Lesson from acute experimental pancreatitis: multidrug strategies is effective than single-target therapy.
PMID 22964964
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G2417
Gabexate mesylate, analytical standard, for drug analysis
C16H23N3O4 · CH4O3S