Annals of biomedical engineering
Analysis of kinetics of dihydroethidium fluorescence with superoxide using xanthine oxidase and hypoxanthine assay.
PMID 22965641
Superoxide (O(2) (-)) is an important reactive oxygen species (ROS), and has an essential role in physiology and pathophysiology. An accurate detection of O(2) (-) is needed to better understand numerous vascular pathologies. In this study, we performed a mechanistic study by using the xanthine oxidase (XOD)/hypoxanthine (HX) assay for O(2) (-) generation and a O(2) (-) sensitive fluorescent dye dihydroethidium (DHE) for O(2) (-) measurement. To quantify O(2) (-) and DHE interactions, we measured fluorescence using a microplate reader. We conducted a detailed reaction kinetic analysis for DHE-O(2) (-) interaction to understand the effect of O(2) (-) self-dismutation and to quantify DHE-O(2) (-) reaction rate. Fluorescence of DHE and 2-hydroethidium (EOH), a product of DHE and O(2) (-) interaction, were dependent on reaction conditions. Kinetic analysis resulted in a reaction rate constant of 2.169xa0±xa00.059xa0×xa010(3)xa0M(-1)xa0s(-1) for DHE-O(2) (-) reaction that is ~100× slower than the reported value of 2.6xa0±xa00.6xa0×xa010(5)xa0M(-1)xa0s(-1). In addition, the O(2) (-) self-dismutation has significant effect on DHE-O(2) (-) interaction. A slower reaction rate of DHE with O(2) (-) is more reasonable for O(2) (-) measurements. In this manner, the DHE is not competing with superoxide dismutase and NO for O(2) (-). Results suggest that an accurate measurement of O(2) (-) production rate may be difficult due to competitive interference for many factors; however O(2) (-) concentration may be quantified.