EMAIL THIS PAGE TO A FRIEND
Journal of immunotoxicology
How aluminum adjuvants could promote and enhance non-target IgE synthesis in a genetically-vulnerable sub-population.
PMID 22967010
Abstract
Aluminum-containing adjuvants increase the effectiveness of vaccination, but their ability to augment immune responsiveness also carries the risk of eliciting non-target responses, especially in genetically susceptible individuals. This study reviews the relevant actions of aluminum adjuvants and sources of genetic risk that can combine to adversely affect a vulnerable sub-population. Aluminum adjuvants promote oxidative stress and increase inflammasome activity, leading to the release of IL-1β, IL-18, and IL-33, but not the important regulatory cytokine IL-12. In addition, they stimulate macrophages to produce PGE₂, which also has a role in regulating immune responses. This aluminum-induced cytokine context leads to a T(H)2 immune response, characterized by the further release of IL-3, IL-4, IL-5, IL-9, IL-13, and IgE-potentiating factors such as sCD23. Genetic variants in cytokine genes, such as IL-4, IL-13, IL-33, and IL-18 influence the response to vaccines in children and are also associated with atopy. These genetic factors may therefore define a genetically-vulnerable sub-population, children with a family history of atopy, who may experience an exaggerated T(H)2 immune response to aluminum-containing vaccines. IL-4, sCD23, and IgE are common factors for both atopy and the immune-stimulating properties of aluminum adjuvants. IL-4 is critical in the production of IgE and total IgE up-regulation. IL-4 has also been reported to induce the production of sCD23 and trigger resting sIgM+, sIgD+ B-cells to switch to sIgE+ B-cells, making them targets for IgE-potentiating factors. Further, the actions of IgE-potentiating factors on sIgE+ B-cells are polyclonal and unrestricted, triggering their differentiation into IgE-forming plasma cells. These actions provide a mechanism for aluminum-adjuvant promotion and enhancement of non-target IgE in a genetically vulnerable sub-population. Identification of these individuals may decrease the risk of adverse events associated with the use of aluminum-containing vaccines.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF81923238
Aluminum, wire reel, 25m, diameter 0.25mm, as drawn, 99.5%
Al
GF42637406
Aluminum, wire reel, 25m, diameter 0.30mm, as drawn, 99.5%
Al
GF34787177
Aluminum, wire reel, 25m, diameter 1.0mm, as drawn, 99.999%
Al
GF35183471
Aluminum, wire reel, 2m, diameter 0.25mm, as drawn, 99.999%
Al
GF47338443
Aluminum, wire reel, 2m, diameter 2.0mm, as drawn, 99.999%
Al
GF51477914
Aluminum, wire reel, 2m, diameter 3.0mm, as drawn, 99.999%
Al
GF20922061
Aluminum, wire reel, 35m, diameter 0.05mm, as drawn, 99.99+%
Al
GF29128855
Aluminum, wire reel, 500m, diameter 0.25mm, as drawn, 99.95+%
Al
GF34949289
Aluminum, wire reel, 500m, diameter 0.5mm, as drawn, 99.95+%
Al
GF88323206
Aluminum, wire reel, 500m, diameter 1.0mm, as drawn, 99.95+%
Al
GF89409673
Aluminum, wire reel, 500m, diameter 0.125mm, as drawn, 99.5%
Al
GF50572155
Aluminum, wire reel, 500m, diameter 0.25mm, as drawn, 99.5%
Al
GF65733958
Aluminum, wire reel, 500m, diameter 0.30mm, as drawn, 99.5%
Al
GF58303098
Aluminum, wire reel, 500m, diameter 0.5mm, as drawn, 99.5%
Al
GF67804117
Aluminum, wire reel, 500m, diameter 0.7mm, as drawn, 99.5%
Al
GF05275403
Aluminum, wire reel, 500m, diameter 1.0mm, as drawn, 99.5%
Al
GF48115313
Aluminum, wire reel, 50m, diameter 0.025mm, as drawn, 99.99+%
Al
GF90308538
Aluminum, wire reel, 50m, diameter 0.125mm, as drawn, 99.99+%
Al
GF22816830
Aluminum, wire reel, 50m, diameter 0.25mm, as drawn, 99.95+%
Al
GF93567921
Aluminum, wire reel, 50m, diameter 0.25mm, hard, 99.99+%
Al
GF75278043
Aluminum, wire reel, 50m, diameter 0.5mm, as drawn, 99.95+%
Al
GF15273929
Aluminum, wire reel, 50m, diameter 1.0mm, as drawn, 99.95+%
Al
GF04280879
Aluminum, wire reel, 50m, diameter 0.063mm, annealed, 99.99%
Al
GF70878976
Aluminum, wire reel, 50m, diameter 0.125mm, as drawn, 99.5%
Al
GF65371327
Aluminum, wire reel, 50m, diameter 0.5mm, as drawn, 99.5%
Al
GF10973550
Aluminum, wire reel, 50m, diameter 0.5mm, as drawn, 99.999%
Al
GF48396730
Aluminum, wire reel, 50m, diameter 1.0mm, as drawn, 99.999%
Al
GF38084430
Aluminum, wire reel, 50m, diameter 1.5mm, as drawn, 99.999%
Al
GF41557035
Aluminum, wire reel, 5m, diameter 0.025mm, as drawn, 99.0+%
Al
GF54185474
Aluminum, wire reel, 5m, diameter 0.025mm, as drawn, 99.99+%
Al