EMAIL THIS PAGE TO A FRIEND
Talanta
Mercury speciation analysis in terrestrial animal tissues.
PMID 22967634
Abstract
No previous analytical procedures are available and validated for mercury speciation analysis in terrestrial animal tissues. This analysis is a difficult task both because the expected concentrations are low, since important accumulation process are not likely to occur, and also because there are not commercially available certified reference material. Thus, an analytical methodology has been developed and validated for mercury speciation for the specific case of terrestrial animal tissues. The proposed method is based on the quantitative extraction of the species by closed-vessel microwave assisted heating with an alkaline reagent, followed by ethylation. The ethylated derivatives were then submitted to head-space solid phase microextraction with a 100 μm polidimethylsiloxane-coated fiber, and desorbed onto a gas chromatograph coupled to atomic fluorescence detection via pyrolysis unit (HS-SPME-GC-pyro-AFS). Procedural detection limits were 31.8 ng g(-1) and 52.5 ng g(-1) for CH(3)Hg(+) and Hg(2+), respectively, for liver and 35.3 ng g(-1) and 58.1 ng g(-1) for CH(3)Hg(+) and Hg(2+), respectively, for kidney. These limits of detection are 5.5 and 6 times better than the obtained without solid phase microextraction for CH(3)Hg(+) and Hg(2+), respectively. The methodology was found linear up to 120 μg L(-1) and reproducible from one day to the following. It was validated with certified reference materials NCS ZC 71001 (beef liver) and BCR No 186 (pig kidney) for total mercury, calculated as the sum of species, and with spiked red deer liver and kidney for speciation. Finally, it was applied to the analysis of samples of red deer liver, red deer kidney and wild boar kidney coming from the Almadén's mercury mining area (Ciudad Real, Spain), the longest and largest producer of mercury in the world until its closure in 2002.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
57300-U SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm, needle size 24 ga, for use with manual holder
57342-U SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm, for use with manual holder, needle size 23 ga
57341-U SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm, for use with autosampler, needle size 23 ga
57928-U SPME fiber assembly Polydimethylsiloxane (PDMS), metal alloy fiber, df 100 μm, needle size 23 ga, for use with autosampler
FFA57301 SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm, for use with multi fiber exchanger, needle size 24 ga
FFA57341-U SPME fiber assembly Polydimethylsiloxane (PDMS), df 100 μm, for use with multi fiber exchanger
504823 SPME Portable Field Sampler, coating Polydimethylsiloxane (PDMS)
87708
Tetramethylammonium bromide, for ion pair chromatography, LiChropur®, ≥99.0% (AT)
C4H12BrN
426296
Tetramethylammonium bromide, ACS reagent, ≥98.0%
C4H12BrN
195758
Tetramethylammonium bromide, 98%
C4H12BrN
87709
Tetramethylammonium bromide, for electrochemical analysis, ≥99.0%
C4H12BrN
87718
Tetramethylammonium chloride, BioUltra, for molecular biology, ≥99.0% (AT)
C4H12ClN
74202
Tetramethylammonium chloride, for ion pair chromatography, LiChropur®, ≥99.0% (AT)
C4H12ClN
T19526
Tetramethylammonium chloride, reagent grade, ≥98%
C4H12ClN
T3411
Tetramethylammonium chloride solution, for molecular biology
459135
Tetramethylammonium fluoride, 97%
C4H12FN
T7505
Tetramethylammonium hydroxide pentahydrate, ≥97%
C4H13NO · 5H2O
87741
Tetramethylammonium hydroxide pentahydrate, ≥95.0% (T)
C4H13NO · 5H2O
426318
Tetramethylammonium hydroxide solution, ACS reagent
C4H13NO
334901
Tetramethylammonium hydroxide solution, 25 wt. % in methanol
C4H13NO
331635
Tetramethylammonium hydroxide solution, 25 wt. % in H2O
C4H13NO
328251
Tetramethylammonium hydroxide solution, 10 wt. % in H2O
C4H13NO
235946
Tetramethylammonium iodide, 99%
C4H12IN
425257
Tetramethylammonium nitrate, 95%
C4H12N2O3
02799
Tetramethylammonium sulfate, for ion pair chromatography, LiChropur®, ≥99.0% (T)
C8H24N2O4S
87743
Tetramethylammonium sulfate hydrate
C8H24N2O4S · xH2O