EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Elastic and bendable caffeine cocrystals: implications for the design of flexible organic materials.
PMID 22969017
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C60007
4-Chloro-3-nitrobenzoic acid, 98%
C7H4ClNO4