EMAIL THIS PAGE TO A FRIEND
Chirality
DNA binding, antioxidant activity, and DNA damage protection of chiral macrocyclic Mn(III) salen complexes.
PMID 22969041
Abstract
We are reporting the synthesis, characterization, and calf thymus DNA binding studies of novel chiral macrocyclic Mn(III) salen complexes S-1, R-1, S-2, and R-2. These chiral complexes showed ability to bind with DNA, where complex S-1 exhibits the highest DNA binding constant 1.20 × 10(6) M(-1). All the compounds were screened for superoxide and hydroxyl radical scavenging activities; among them, complex S-1 exhibited significant activity with IC(50) 1.36 and 2.37 μM, respectively. Further, comet assay was used to evaluate the DNA damage protection in white blood cells against the reactive oxygen species wherein complex S-1 was found effective in protecting the hydroxyl radicals mediated plasmid and white blood cells DNA damage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0742 EUK-8, ≥95% (HPLC)
C16H14ClMnN2O2