EMAIL THIS PAGE TO A FRIEND
Nederlands tijdschrift voor geneeskunde
[A man with progressive diplopia].
PMID 22971425
Abstract
A 71-year-old man presented with progressive diplopia due to swelling of a Miragel explant. Removal of the explant resulted in relief of symptoms.