EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Novel coumarin derivatives containing 1,2,4-triazole, 4,5-dicyanoimidazole and purine moieties: synthesis and evaluation of their cytostatic activity.
PMID 22971585
Abstract
We report here on the synthesis and in vitro anti-tumor effects of a series of novel 1,2,4-triazole (compounds 3-6), 4,5-dicyanoimidazole (compound 7), and purine (compounds 8-13) coumarin derivatives and their acyclic nucleoside analogues 14-18. Structures of novel compounds 3-18 were deduced from their (1)H- and (13)C-NMR and corresponding mass spectra. Results of anti-proliferative assays performed on a panel of selected human tumor cell lines revealed that compound 6 had moderate cytostatic activity against the HeLa cell line (IC(50) = 35 µM), whereas compound 10 showed moderate activity against the HeLa (IC(50) = 33 µM), HepG2 (IC(50) = 25 µM) and SW620 (IC(50) = 35 µM) cell lines. These compounds showed no cytotoxic effects on normal (diploid) human fibroblasts.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T46108
1,2,4-Triazole, 98%
C2H3N3