EMAIL THIS PAGE TO A FRIEND
Critical care medicine
Etomidate is associated with mortality and adrenal insufficiency in sepsis: a meta-analysis*.
PMID 22971586
Abstract
To evaluate the effects of single-dose etomidate on the adrenal axis and mortality in patients with severe sepsis and septic shock. A systematic review of randomized controlled trials and observational studies with meta-analysis. Literature search of EMBASE, Medline, Cochrane Database, and Evidence-Based Medical Reviews. Sepsis patients who received etomidate for rapid sequence intubation. None. We conducted a systematic review of randomized controlled trials and observational studies with meta-analysis assessing the effects of etomidate on adrenal insufficiency and all-cause mortality published between January 1950 and February 2012. We only examined studies including septic patients. All-cause mortality served as our primary end point, whereas the prevalence of adrenal insufficiency was our secondary end point. Adrenal insufficiency was determined using a cosyntropin stimulation test in all studies. We used a random effects model for analysis; heterogeneity was assessed with the I statistic. Publication bias was evaluated with Begg's test. Five studies were identified that assessed mortality in those who received etomidate. A total of 865 subjects were included. Subjects who received etomidate were more likely to die (pooled relative risk 1.20; 95% confidence interval 1.02-1.42; Q statistic, 4.20; I2 statistic, 4.9%). Seven studies addressed the development of adrenal suppression associated with the administration of etomidate; 1,303 subjects were included. Etomidate administration increased the likelihood of developing adrenal insufficiency (pooled relative risk 1.33; 95% confidence interval 1.22-1.46; Q statistic, 10.7; I2 statistic, 43.9%). Administration of etomidate for rapid sequence intubation is associated with higher rates of adrenal insufficiency and mortality in patients with sepsis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E6530
Etomidate, >98% (HPLC), powder
C14H16N2O2