EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Accumulation of ACE inhibitory tripeptides, Val-Pro-Pro and Ile-Pro-Pro, in vascular endothelial cells.
PMID 22972320
Abstract
The antihypertensive peptides, Val-Pro-Pro and Ile-Pro-Pro, were successfully detected in the aorta of spontaneously hypertensive rats after orally administering these peptides by a guanidine-thiocyanate treatment to prevent proteolysis. Cy3-labeled versions of both peptides were localized in the endothelial cells of arterial vessels in the rats. The accumulation of both peptides in the endothelial cells suggested in vivo inhibitory activity of the angiotensin I-converting enzyme.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50983
Guanidine thiocyanate solution, BioUltra, for molecular biology, ~6 M in H2O
CH5N3 · CHNS