EMAIL THIS PAGE TO A FRIEND
Clinical orthopaedics and related research
All-polyethylene tibial components are equal to metal-backed components: systematic review and meta-regression.
PMID 22972656
Abstract
Less than 1% of all primary TKAs are performed with an all-polyethylene tibial component, although recent studies indicate all-polyethylene tibial components are equal to or better than metal-backed ones. We asked whether the metal-backed tibial component was clinically superior to the all-polyethylene tibial component in primary TKAs regarding revision rates and clinical functioning, and which modifying variables affected the revision rate. We systematically reviewed the literature for clinical studies comparing all-polyethylene and metal-backed tibial components used in primary TKAs in terms of revision rates, clinical scores, and radiologic parameters including radiostereometric analysis (RSA). Meta-regression techniques were used to explore factors modifying the observed effect. Our search yielded 1557 unique references of which 26 articles were included, comprising more than 12,500 TKAs with 231 revisions for any reason. Meta-analysis showed no differences between the all-polyethylene and metal-backed components except for higher migration of the metal-backed components. Meta-regression showed strong evidence that the all-polyethylene design has improved with time compared with the metal-backed design. The all-polyethylene components were equivalent to metal-backed components regarding revision rates and clinical scores. The all-polyethylene components had better fixation (RSA) than the metal-backed components. The belief that metal-backed components are better than all-polyethylene ones seems to be based on studies from earlier TKAs. This might no longer be true for modern TKAs. Level II, therapeutic study. See Guidelines for Authors for a complete description of levels of evidence.