EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
B-(3,3-difluoroallyl)diisopinocampheylborane for the enantioselective fluoroallylboration of aldehydes.
PMID 22973971
Abstract
The fluoroallylboration of aldehydes with B-(3,3-difluoroallyl)diisopinocampheylborane, which was prepared via the hydroboration of 1,1-difluoroallene, provides chiral 2,2-gem-difluorinated homoallylic alcohols in good yields and 91-97% ee.
Related Materials