EMAIL THIS PAGE TO A FRIEND
Journal of colloid and interface science
Embossing of organic thin films using a surfactant assisted lift-off technique.
PMID 22975399
Abstract
A simple technique for patterning organic materials using a surfactant assisted lift-off method is proposed. Thin films of various organic materials are prepared, and areas in contact with a surfactant coated poly(dimethylsiloxane) (PDMS) stamp are selectively removed. The general applicability of this technique is shown for materials containing nitrate, amine, and carboxylic acid functional groups. This technique provides a new methodology for fabricating patterns with vertical dimensions ranging from 30 nm up to 3 μm on organic thin films with specific functional groups.