EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Polycationic gramicidin S analogues with both high antibiotic activity and very low hemolytic activity.
PMID 22976321
Abstract
The substitution of each constituent amino acid residue of gramicidin S (GS), cyclo(-Val(1,1')-Orn(2,2')-Leu(3,3')-D-Phe(4,4')-Pro(5,5')-)(2) with Lys residue indicated that each side chain structure of the constituent amino acid residues affect largely the antibiotic activity and hemolytic activity of GS. Further, the substitution of D-Phe(4,4') and Pro(5,5') residues with basic amino acid residues as a Lys residue results the high antibiotic activity and the very low hemolytic activity. Thus, we have found novel positions on the scaffold of GS at D-Phe(4,4') and Pro(5,5') residues whose modification will significantly increase the therapeutic index.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
50845
Gramicidin A from Bacillus brevis