EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
Synthesis and evaluation of novel 3-C-alkylated-Neu5Ac2en derivatives as probes of influenza virus sialidase 150-loop flexibility.
PMID 22976385
Abstract
Novel 3-C-alkylated-Neu5Ac2en derivatives have been designed to target the expanded active site cavity of influenza virus sialidases with an open 150-loop, currently seen in X-ray crystal structures of influenza A virus group-1 (N1, N4, N5, N8), but not group-2 (N2, N9), sialidases. The compounds show selectivity for inhibition of H5N1 and pdm09 H1N1 sialidases over an N2 sialidase, providing evidence of the relative 150-loop flexibility of these sialidases. In a complex with N8 sialidase, the C3 substituent of 3-phenylally-Neu5Ac2en occupies the 150-cavity while the central ring and the remaining substituents bind the active site as seen for the unsubstituted template. This new class of inhibitors, which can 'trap' the open 150-loop form of the sialidase, should prove useful as probes of 150-loop flexibility.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9050
N-Acetyl-2,3-dehydro-2-deoxyneuraminic acid, ≥93%
C11H17NO8