EMAIL THIS PAGE TO A FRIEND
Food chemistry
Quantitative determination of the boar taint compounds androstenone, skatole, indole, 3α-androstenol and 3β-androstenol in wild boars (Sus scrofa) reveals extremely low levels of the tryptophan-related degradation products.
PMID 22980780
Abstract
The major boar taint compounds androstenone and skatole as well as the minor compounds indole, 3α-androstenol and 3β-androstenol were determined in back fat samples of 23 male wild boars by applying a recently published SIDA-HS-SPME-GC/MS method. The boar pheromones androstenone, 3α-androstenol and 3β-androstenol were found in extraordinary high concentrations, resulting in mean values of 3329ng/g androstenone, 1273 ng/g 3α-androstenol and 545 ng/g 3β-androstenol. Interestingly, skatole was not detectable in about 50% of the boars and negligibly low in all other samples as expressed by a mean skatole value of only 14 ng/g. Indole was also found in every sample, but again in low concentrations with a mean value of 40 ng/g. Possible factors explaining this remarkably low skatole deposition in wild boars such as intestinal flora and anatomy, dietary composition, housing or genetic predisposition are discussed in this paper.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M51458
3-Methylindole, 98%
C9H9N
W301912
Skatole, ≥98%
C9H9N