EMAIL THIS PAGE TO A FRIEND
Neurotoxicology
Blood harmane (1-methyl-9H-pyrido[3,4-b]indole) concentration in essential tremor cases in Spain.
PMID 22981972
Abstract
Environmental correlates for essential tremor (ET) are largely unexplored. The search for such environmental factors has involved the study of a number of neurotoxins. Harmane (1-methyl-9H-pyrido[3,4-b]indole) is a potent tremor-producing toxin. In two prior case-control studies in New York, we demonstrated that blood harmane concentration was elevated in ET patients vs. controls, and especially in familial ET cases. These findings, however, have been derived from a study of cases ascertained through a single tertiary referral center in New York. Our objective was to determine whether blood harmane concentrations are elevated in familial and sporadic ET cases, ascertained from central Spain, compared to controls without ET. Blood harmane concentrations were quantified by a well-established high performance liquid chromatography method. The median harmane concentrations were: 2.09 g(-10)/ml (138 controls), 2.41 g(-10)/ml (68 sporadic ET), and 2.90 g(-10)/ml (62 familial ET). In an unadjusted logistic regression analysis, log blood harmane concentration was not significantly associated with diagnosis (familial ET vs. control): odds ratio=1.56, p=0.26. In a logistic regression analysis that adjusted for evaluation start time, which was an important confounding variable, the odds ratio increased to 2.35, p=0.049. Blood harmane levels were slightly elevated in a group of familial ET cases compared to a group of controls in Spain. These data seem to further extend our observations from New York to a second cohort of ET cases in Spain. This neurotoxin continues to be a source of interest for future confirmatory research.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
103276
Harmane, 98%
C12H10N2
286044
Harmine, 98%
C13H12N2O