EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Reaction of glycal derivatives with alcohols in the presence of N-bromosuccinimide and diphenyldiselenide: preparation of 2-deoxy-2-phenylselenyl glycosides.
PMID 22982170
Abstract
Reaction of glycal derivatives with alcohols or glycosyl acceptors in the presence of N-bromosuccinimide (NBS) and diphenyldiselenide resulted in the formation of alkyl 2-deoxy-2-phenylselenyl glycosides or disaccharide derivatives in excellent yield. The reactions are reasonably fast and considerable stereo-selectivity was observed in the preparation of disaccharide derivatives.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42944
Diphenyl diselenide, purum, ≥97.0% (GC)
C12H10Se2
180629
Diphenyl diselenide, 98%
C12H10Se2