EMAIL THIS PAGE TO A FRIEND
Infection
Antimicrobial prescribing in nursing homes in Finland: results of three point prevalence surveys.
PMID 22983808
Abstract
The purpose of this study was to analyse the Finnish European Surveillance of Antimicrobial Consumption (ESAC) nursing home (NH) point prevalence surveys' (PPSs) data in detail, i.e. to evaluate the variability in the prevalence of antimicrobial prescription between NHs and its relationship to resident characteristics. All residents present in NHs for ≥ 24 h and receiving systemic antimicrobials on the day of the survey were included. Data on antimicrobials and their indications (prophylaxis or treatment, type of infection) were collected. Three PPSs were performed: eight NHs participated in April and November 2009 and nine in May-September 2010. In total, there were 5,691 eligible residents (range by survey, 1,706-2,320; range by NH, 60-688), 716 (12.6 %; range by NH, 3.2-33.3 %) of which received at least one antimicrobial and 40 residents received two. The most common indication was prophylaxis (487/5,691, 8.6 %), mainly for urinary tract infection (UTI) (460/487, 94.5 %). Of the residents, 269/5,691 (4.7 %, range by NH, 1.5-6.0 %) were on antimicrobial treatment. UTI (119/269; 44.2 %) was the most common indication for treatment. Methenamine (306/756, 40.5 %) was the most commonly used antimicrobial, followed by trimethoprim (13.6 %) and pivmecillinam (11.0 %). In the eight NHs participating in all three surveys, the prevalence of residents receiving antimicrobials decreased from 16.6 to 9.7 %. Antimicrobial use was common in NHs in Finland and most were used for UTI prophylaxis and treatment. The usage, however, varied among NHs and tended to decrease during the surveys. NHs may benefit from antimicrobial stewardship interventions focused on UTI.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0817
Pivmecillinam, ≥98% (HPLC)
C21H33N3O5S