EMAIL THIS PAGE TO A FRIEND
Organic letters
Glycosylations of cyclopropyl-modified carbohydrates: remarkable β-selectivity using a mannose building block.
PMID 22985402
Abstract
A mannosyl donor bearing a spiroannulated cyclopropane unit at C-5 has been prepared, and its behavior in glycosylation reactions investigated. Selectivities in favor of the β-anomer were observed. Corresponding di- and trisaccharides incorporating the rigid cyclopropane motif were assembled.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
295183
Cyclopropane, ≥99%
C3H6