EMAIL THIS PAGE TO A FRIEND
Journal of the American Veterinary Medical Association
Waiting for the Food and Drug Administration to respond to its advisory committee's recommendations.
PMID 2298647
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32173
Ipronidazole, VETRANAL, analytical standard
C7H11N3O2