EMAIL THIS PAGE TO A FRIEND
Journal of the American Veterinary Medical Association
Drug-associated aplastic anemia in dogs: eight cases (1984-1988).
PMID 2298681
Abstract
Records of 8 dogs with drug-associated aplastic anemia were reviewed. Drugs suspected as being causative included estradiol cyclopentylpropionate (3 dogs), phenylbutazone (2 dogs), meclofenamic acid (1 dog), trimethoprim-sulfadiazine and fenbendazole (1 dog), and quinidine (1 dog). Five of the dogs died or were euthanatized. One dog with estrogen-associated aplasia recovered after prolonged treatment. The dogs with trimethoprim-sulfadiazine and quinidine-associated marrow aplasia recovered promptly after treatment was discontinued.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Q5001
Quinidine gluconate salt, analytical standard
C20H24N2O2 · C6H12O7