EMAIL THIS PAGE TO A FRIEND
Cutis
Randomized, observer-blind, split-face study to compare the irritation potential of 2 topical acne formulations over a 14-day treatment period.
PMID 22988653
Abstract
This randomized, observer-blind, split-face study assessed the irritation potential and likelihood of continued use of clindamycin phosphate 1.2%--benzoyl peroxide (BPO) 2.5% gel or adapalene 0.1%--BPO 2.5% gel once daily over a 14-day treatment period in 21 participants (11 males; 10 females) with acne who were 18 years or older. Investigator clinical assessment (erythema and dryness) and self-assessment (dryness and burning/stinging) were performed at baseline and each study visit (days 1-14) using a 4-point scale (O = none; 3 = severe). Transepidermal water loss (TEWL) and corneometry measurements were performed at baseline and days 3, 5, 7, 9, 11, and 14. Lesions were counted at baseline and on day 14. Participant satisfaction questionnaires were completed on days 7 and 14. At the end of the study, investigators reported none or only mild erythema in 86% (18/21) of participants treated with clindamycin phosphate 1.2%--BPO 2.5% gel compared with 62% (13/21) of participants treated with adapalene 0.1%--BPO 2.5% gel. No severe erythema was reported with clindamycin phosphate 1.2%--BPO 2.5% gel. Adapalene 0.1%--BPO 2.5% gel was prematurely discontinued due to severe erythema in 1 participant on day 5 and a second participant on day 9. Additionally, 2 more participants reported severe erythema on day 14. Mean erythema scores were 0.9 (mean change from baseline, 0.7) with clindamycin phosphate 1.2%--BPO 2.5% gel and 1.4 (mean change from baseline, 1.3) with adapalene 0. 1%--BPO 2.5% gel on day 14 (P < .05 for days 6-14). Similar results were seen with dryness. Mean scores were 0.5 (mean change from baseline, 0.4) and 1.0 (mean change from baseline, 1.0), respectively (P < .05 for days 6-14). Self-assessment, TEWL, and corneometry results underscored the investigator clinical assessment. Participant preference and likelihood of continued usage was greater with clindamycin phosphate 1.2%--BPO 2.5% gel. Continued use and efficacy results for the treatment of acne were influenced by the potential of the product to cause irritation and the participant preferences. Irritation potential was more pronounced and severe with adapalene 0.1%--BPO 2.5% gel. Undoubtedly, as a result more participants preferred treatment with clindamycin phosphate 1.2%--BPO 2.5% gel and were more likely to continue to use the product.