EMAIL THIS PAGE TO A FRIEND
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.]
A patient with Bowen's disease successfully treated using a 1,927-nm thulium fiber fractional laser.
PMID 22988995