EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Solvent effect in reactions using Stryker's reagent.
PMID 22989178
Abstract
The solvent has a significant influence in the rate of reactions promoted by Stryker's reagent. The reactions performed in THF were, in most cases, faster than in toluene.