EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Letter to the Editor on "Synthetic cyclohexenyl chalcone natural products possess cytotoxic activities against prostate cancer cells and inhibit cysteine cathepsins in vitro".
PMID 22989751
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11970
1,3-Diphenyl-2-propenone, ≥98.0% (GC)
C15H12O