EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
AlCl3 mediated unexpected migration of sulfonyl groups: regioselective synthesis of 7-sulfonyl indoles of potential pharmacological interest.
PMID 22990283
Abstract
A conceptually new and straightforward introduction of sulfonyl groups at the C-7 position of an indole ring has been achieved via AlCl(3) mediated unexpected regioselective sulfonyl group migration for N-alkyl/aryl/heteroarylsulfonyl indoles affording potential inhibitors of Mycobacterium tuberculosis H37Rv chorismate mutase.