EMAIL THIS PAGE TO A FRIEND
Journal of hypertension
Higher cardiovascular risk and impaired benefit of antihypertensive treatment in hypertensive patients requiring additional drugs on top of randomized therapy: is adding drugs always beneficial?
PMID 22990352
Abstract
In antihypertensive treatment trials, when randomized therapies do not reach target, additional drugs are administered. However, patients requiring (add-on) or not requiring add-on therapy (no-add-on) may be at different cardiovascular risk and differently susceptible to benefits of antihypertensive treatment. The Felodipine Event Reduction study included 9711 Chinese hypertensive patients receiving 12.5  mg/day hydrochlorothiazide and randomized to associating either felodipine (5  mg/day) or placebo. Within 6 months, add-on therapy (further diuretic and other drugs) was required by 2185 patients, whereas 7243 did not require it. Despite significant SBP/DBP reductions by add-on therapy, outcome incidence remained much lower in no-add-on than in add-on patients: hazard ratios for various outcomes, after adjusting for baseline variables and blood pressure (BP) at time of add-on decision, were 0.22-0.368 (P always <0.001) and remained substantially unchanged when further adjusted for the small SBP/DBP difference persisting during follow-up treatment (-2.4/-1.1  mmHg in no-add-on). When felodipine was compared to placebo, the benefit of a lower SBP/DBP caused by felodipine was evident in the no-add-on patients (hazard ratio 0.45-0.68, P always <0.001), but it was lost in the add-on group (hazard ratio 0.91-1.17). Comparing patients more or less easily responding to antihypertensive treatment may identify patients at high risk of outcomes and less susceptible to benefits of a lower BP. It remains to be more directly investigated to what extent adding drugs to drugs is effective in reducing outcomes of patients in whom simple antihypertensive therapy does not achieve goal BP.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F9677
Felodipine, solid
C18H19NO4Cl2