EMAIL THIS PAGE TO A FRIEND
Cancer
UBE4B levels are correlated with clinical outcomes in neuroblastoma patients and with altered neuroblastoma cell proliferation and sensitivity to epidermal growth factor receptor inhibitors.
PMID 22990745
Abstract
The UBE4B gene, which is located on chromosome 1p36, encodes a ubiquitin ligase that interacts with hepatocyte growth factor-regulated tyrosine kinase substrate (Hrs), a protein involved in epidermal growth factor receptor (EGFR) trafficking, suggesting a link between EGFR trafficking and neuroblastoma pathogenesis. The authors analyzed the roles of UBE4B in the outcomes of patients with neuroblastoma and in neuroblastoma tumor cell proliferation, EGFR trafficking, and response to EGFR inhibition. The association between UBE4B expression and the survival of patients with neuroblastoma was examined using available microarray data sets. UBE4B and EGFR protein levels were measured in patient tumor samples, EGFR degradation rates were measured in neuroblastoma cell lines, and the effects of UBE4B on neuroblastoma tumor cell growth were analyzed. The effects of the EGFR inhibitor cetuximab were examined in neuroblastoma cells that expressed wild-type and mutant UBE4B. Low UBE4B gene expression is associated with poor outcomes in patients with neuroblastoma. UBE4B overexpression reduced neuroblastoma tumor cell proliferation, and UBE4B expression was inversely related to EGFR expression in tumor samples. EGFR degradation rates correlated with cellular UBE4B levels. Enhanced expression of catalytically active UBE4B resulted in reduced sensitivity to EGFR inhibition. The current study demonstrates associations between UBE4B expression and the outcomes of patients with neuroblastoma and between UBE4B and EGFR expression in neuroblastoma tumor samples. Moreover, levels of UBE4B influence neuroblastoma tumor cell proliferation, EGFR degradation, and response to EGFR inhibition. These results suggest UBE4B-mediated growth factor receptor trafficking may contribute to the poor prognosis of patients who have neuroblastoma tumors with 1p36 deletions and that UBE4B expression may be a marker that can predict responses of neuroblastoma tumors to treatment.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV54552
Anti-UBE4B antibody produced in rabbit, affinity isolated antibody