EMAIL THIS PAGE TO A FRIEND
Environmental science & technology
Reductive dissolution of Tl(I)-jarosite by Shewanella putrefaciens: providing new insights into Tl biogeochemistry.
PMID 22992155
Abstract
Thallium (Tl) is emerging as a metal of concern in countries such as China due to its release during the natural weathering of Tl-bearing ore deposits and mining activities. Despite the high toxicity of Tl, few studies have examined the reductive dissolution of Tl mineral phases by microbial populations. In this study we examined the dissolution of synthetic Tl(I)-jarosite, (H(3)O)(0.29)Tl(0.71)Fe(2.74)(SO(4))(2)(OH)(5.22)(H(2)O)(0.78), by Shewanella putrefaciens CN32 using batch experiments under anaerobic circumneutral conditions. Fe(II) concentrations were measured over time and showed Fe(II) production (4.6 mM) in inoculated samples by 893 h not seen in mineral and dead cell controls. Release of aqueous Tl was enhanced in inoculated samples whereby maximum concentrations in inoculated and cell-free samples reached 3.2 and 2.1 mM, respectively, by termination of the experiment. Complementary batch Tl/S. putrefaciens sorption experiments were conducted under experimentally relevant pH (5 and 6.3) at a Tl concentration of 35 μM and did not show significant Tl accumulation by either live or dead cells. Therefore, in contrast to many metals such as Pb and Cd, S. putrefaciens does not represent a sink for Tl in the environment and Tl is readily released from Tl-jarosite during both abiotic and biotic dissolution.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
277932
Thallium, granular, 1-5 mm diameter, 99.99% trace metals basis
Tl